Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD307d (FcRL4)
Supplier:  Biolegend 340211
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD307d (FcRL4) [413D12]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-6430