Learn More
Biolegend PerCP/Cyanine5.5 anti-human/mouse CXCR7
Supplier:  Biolegend 331120
Encompass_Preferred
PerCP/Cyanine5.5 anti-human/mouse CXCR7 [8F11-M16]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: FC; Size: 100 tests
Catalog No. 50-207-5989