Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD99
Supplier:  Biolegend 398210
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD99 [hec2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9358