Learn More
MSC Solenis Cleaner, Hydrogen Peroxide, Concentrated, 4/Pk.
Catalog No. 502817376
Encompass
This concentrated cleaner utilizes proprietary Accelerated Hydrogen Peroxide (AHP®) technology, providing a heavy-duty solution safe for various hard surfaces, including marble. It is versatile for applications like floors, walls, and glass, and can be used for carpet extraction, prespray, and spotting.
  • Uses patented Accelerated Hydrogen Peroxide (AHP®) technology for effective cleaning
  • Heavy-duty cleaner safe on most hard surfaces, including marble floors
  • Concentrated formula offers cost control and excellent cleaning performance
  • Phosphorous-free with no added dyes or fragrance
  • Suitable for autoscrubbers, mop and bucket, and spray-and-wipe applications
  • Can be used for carpet spotting and extraction cleaning
Catalog No. 50-281-7376 Supplier MSC Supplier No. 02985992
This concentrated cleaner utilizes proprietary Accelerated Hydrogen Peroxide (AHP®) technology, providing a heavy-duty solution safe for various hard surfaces, including marble. It is versatile for applications like floors, walls, and glass, and can be used for carpet extraction, prespray, and spotting.
  • Uses patented Accelerated Hydrogen Peroxide (AHP®) technology for effective cleaning
  • Heavy-duty cleaner safe on most hard surfaces, including marble floors
  • Concentrated formula offers cost control and excellent cleaning performance
  • Phosphorous-free with no added dyes or fragrance
  • Suitable for autoscrubbers, mop and bucket, and spray-and-wipe applications
  • Can be used for carpet spotting and extraction cleaning