Learn More
MSC Diversey PERdiem General Purpose Cleaner, Hydrogen Peroxide, 1.5 gal
Catalog No. 502817354
Encompass
PERdiem General Purpose Cleaner with Hydrogen Peroxide is a concentrated liquid cleaning product designed for industrial and institutional applications. This clear, colorless, and odorless cleaner is intended for dilution prior to use, providing an effective and safe cleaning solution.
  • Formulated with hydrogen peroxide for powerful cleaning
  • Concentrated solution for industrial and institutional use
  • Undiluted pH of less than 2, approximately 6.2 when diluted
  • Completely soluble in water for easy mixing
  • Non-corrosive to metals
  • VOC content of 0%
Catalog No. 50-281-7354 Supplier MSC Supplier No. 03007887
PERdiem General Purpose Cleaner with Hydrogen Peroxide is a concentrated liquid cleaning product designed for industrial and institutional applications. This clear, colorless, and odorless cleaner is intended for dilution prior to use, providing an effective and safe cleaning solution.
  • Formulated with hydrogen peroxide for powerful cleaning
  • Concentrated solution for industrial and institutional use
  • Undiluted pH of less than 2, approximately 6.2 when diluted
  • Completely soluble in water for easy mixing
  • Non-corrosive to metals
  • VOC content of 0%