Learn More
MSC Diversey All-Purpose Cleaner, Disinfectant, Hydrogen Peroxide Formula, Fragrance-Free, 1 gal
Catalog No. 502818842
Encompass
This all-purpose cleaner comes in a 1 gal bottle and functions as a disinfectant. It is a liquid with a fragrance-free scent, formulated with hydrogen peroxide.
  • Liquid form
  • Fragrance-free scent
  • Disinfectant capabilities
  • Hydrogen peroxide formula
  • Certified by Ecologo, Green Seal, Halal, and Kosher
  • Effective against dust, germs, grease, mildew, mold, and various soils
  • Suitable for cleaning sealed wood, vinyl, laminate, fiberglass, enamel, glass, finished wood, plastic, metal, engineered stone, sealed stone, ceramic, and grout
Catalog No. 50-281-8842 Supplier MSC Supplier No. 10725679
This all-purpose cleaner comes in a 1 gal bottle and functions as a disinfectant. It is a liquid with a fragrance-free scent, formulated with hydrogen peroxide.
  • Liquid form
  • Fragrance-free scent
  • Disinfectant capabilities
  • Hydrogen peroxide formula
  • Certified by Ecologo, Green Seal, Halal, and Kosher
  • Effective against dust, germs, grease, mildew, mold, and various soils
  • Suitable for cleaning sealed wood, vinyl, laminate, fiberglass, enamel, glass, finished wood, plastic, metal, engineered stone, sealed stone, ceramic, and grout