Learn More
Edge Biosystems Inc PERF. DTR ULTRA 96-WELL PLT KT
SDP
Catalog No. NC9660137
Encompass
fully hydrated gel filtration matrix that provides for a convenient and simple method of dye terminator removal for sequencing cleanup.
Catalog No. NC9660137 Supplier Edge Biosystems Inc Supplier No. 54789
fully hydrated gel filtration matrix that provides for a convenient and simple method of dye terminator removal for sequencing cleanup.