Learn More
Electron Microscopy Sciences Perfect Loop, Loop only, 5 each
SDP
Catalog No. 50930560
Encompass_Preferred
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases. For Ultra Thin sections.
Catalog No. 50-930-560 Supplier Electron Microscopy Sciences Supplier No. 70946
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases. For Ultra Thin sections.