Learn More
Electron Microscopy Sciences Perfect Loop, Set of Handle & Loop
SDP
Catalog No. 50930558
Encompass_Preferred
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases. Set includes loop and handle.
Catalog No. 50-930-558 Supplier Electron Microscopy Sciences Supplier No. 70944
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases. Set includes loop and handle.