Learn More
Agilent Technologies PERFECT MATCH PCR ENHANCER
Catalog No. 50125912
Encompass
Perfect Match PCR Enhancer 100 U Increases yield specificity of primary PCR amplification products Minimizes formation of poorly matched primer-template complexes Add to genomic reactions to destabilize
This item is not returnable. View return policy
Catalog No. 50-125-912 Supplier Agilent Technologies Supplier No. 600129
Perfect Match PCR Enhancer 100 U Increases yield specificity of primary PCR amplification products Minimizes formation of poorly matched primer-template complexes Add to genomic reactions to destabilize