Learn More
Agilent Technologies PERFECT MATCH PCR ENHANCER
Supplier:  Agilent Technologies 600130
Encompass
Perfect Match PCR Enhancer 200 U Increases yield and specificity of primary PCR amplification products Minimizes formation of poorly matched primer-template complexes Add to genomic PCR reactions to destabilize
Catalog No. 50-125-913