Learn More
Electron Microscopy Sciences Perfect Scale Only
SDP
Catalog No. 50930413
Encompass_Preferred
Made from Nickel alloy, 3 mm in length with 1/10 mm divisions. The Scale fits the Perfect Handle (70616-HD). The Perfect Scale comes complete with one scale and a handle.
Catalog No. 50-930-413 Supplier Electron Microscopy Sciences Supplier No. 7061720
Made from Nickel alloy, 3 mm in length with 1/10 mm divisions. The Scale fits the Perfect Handle (70616-HD). The Perfect Scale comes complete with one scale and a handle.