Learn More
Genhunter Corporation PerfectFilm, 8x10 in, 100 sheets
Supplier:  Genhunter Corporation B581
Encompass
•Standard High Contrast, Double Emulsion Blue Film •High Sensitivity With Low Background •Optimized For Ecl (Enhanced Chemiluminescence), But Ideal For Many Other Applications (See Below) •Beautiful, Publication Quality Images •Also Works Well With 14C, 32P, 33P, 35S, 125I 8 X 10 Inches (20.3 X 25.4 Cm) 100 Sheets.
Catalog No. NC0529809