Learn More
Sigma Aldrich Fine Chemicals Biosciences PerfectHybTM Plus Hybrdz125ML
Catalog No. 502901543
Encompass
Sigmas PerfectHyb Plus is a ready-to-use hybridization buffer for Northern and Southern blot protocols. Use of PerfectHyb Plus can decrease hybridization time to as little as two hours.
Catalog No. 50-290-1543 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H7033125ML
Sigmas PerfectHyb Plus is a ready-to-use hybridization buffer for Northern and Southern blot protocols. Use of PerfectHyb Plus can decrease hybridization time to as little as two hours.