Learn More
Sigma Aldrich Fine Chemicals Biosciences PerfectHybTM Plus Hybridiz1L
Catalog No. 502914187
Encompass
Sigmas PerfectHyb Plus is a ready-to-use hybridization buffer for Northern and Southern blot protocols. Use of PerfectHyb Plus can decrease hybridization time to as little as two hours.
Catalog No. 50-291-4187 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H70331L
Sigmas PerfectHyb Plus is a ready-to-use hybridization buffer for Northern and Southern blot protocols. Use of PerfectHyb Plus can decrease hybridization time to as little as two hours.