Learn More
MSC Perfection Chain Products Single Jack Chain, 0.135 in Diameter, Steel, Pre-Galvanized
Catalog No. 5000537384
Encompass
This single jack chain is constructed from steel with a pre-galvanized finish, featuring a 0.135 inch diameter. It is a type of weldless chain made with thin wire and figure-eight-shaped links, suitable for various general utility applications.
  • Constructed from durable steel
  • Features a pre-galvanized finish for corrosion resistance
  • Weldless chain design with figure-eight-shaped links
  • Suitable for hanging light fixtures, signs, and novelties
  • Offers a work load limit of 43 lb
  • Available in a total coil length of 100 feet
Catalog No. 50-005-37384 Supplier MSC Supplier No. 67775296
This single jack chain is constructed from steel with a pre-galvanized finish, featuring a 0.135 inch diameter. It is a type of weldless chain made with thin wire and figure-eight-shaped links, suitable for various general utility applications.
  • Constructed from durable steel
  • Features a pre-galvanized finish for corrosion resistance
  • Weldless chain design with figure-eight-shaped links
  • Suitable for hanging light fixtures, signs, and novelties
  • Offers a work load limit of 43 lb
  • Available in a total coil length of 100 feet