Learn More
MSC Perfection Chain Single Jack Chain, Steel, 0.062 inch Diameter, Zinc Plated
Catalog No. 5000527368
Encompass
Perfection Chain Single Jack Chain is a durable, zinc plated chain made from steel, featuring a 0.062 inch diameter. This chain is designed for a variety of applications where a robust, non-lifting chain is required, and is supplied in a 500 feet total coil length.
  • Zinc plated finish for enhanced durability
  • Not suitable for lifting applications
  • Overall length of 500 feet
  • 0.520 inch inside length
  • 23 links per foot
  • Work load limit of 10 lb
Catalog No. 50-005-27368 Supplier MSC Supplier No. 67775247
Perfection Chain Single Jack Chain is a durable, zinc plated chain made from steel, featuring a 0.062 inch diameter. This chain is designed for a variety of applications where a robust, non-lifting chain is required, and is supplied in a 500 feet total coil length.
  • Zinc plated finish for enhanced durability
  • Not suitable for lifting applications
  • Overall length of 500 feet
  • 0.520 inch inside length
  • 23 links per foot
  • Work load limit of 10 lb