Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ Lite-N-Tite Angled Upright Brooms

Catalog No. 19042013C Shop All Perfex Corp. Products
Change view
Click to view available options
Color:
Blue
Red
White
Description:
Flagged end
Unflagged end
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Description
19-042-013C White Flagged end
19-042-013A Blue Flagged end
19-042-014C White Unflagged end
19-042-013B Red Flagged end
19-042-014A Blue Unflagged end
19-042-014B Red Unflagged end
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-042-013C Supplier Perfex™ Supplier No. 2200W
Add to Cart
Add to Cart

Requires

Requires 19-042-036 or 19-042-041 handle.

Provides thorough sweeping with a single pass. Angled polypropylene fibers provide uniform surface contact, even when surfaces are irregular. Polypropylene fibers are fused onto a high-impact resistant polymer block, ensuring long-lasting durability and no absorption of odors, liquids, or bacteria. Available with a flagged end for fine sweeping, or with an unflagged end for coarse sweeping and scrubbing on surfaces like concrete, tile, and asphalt. A patented self-locking connection system prevents handle breakage and keeps broom securely attached. Choice of blue, red, and white.

Specifications

Product Type Angled Upright Brush
Color White
Description Flagged end
For Use With (Application) A wide range of cleaning requirements with Perfex Lite-N-Tite upright brooms
For Use With (Equipment) Adjustable Polymer Handle
Material Polypropylene Fiber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.