Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ Polymer Fixed Length Handle

Catalog No. 19142742 Shop All Perfex Corp. Products
Change view
Click to view available options
Material:
Polymer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-142-742 Polymer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-142-742 Supplier Perfex™ Supplier No. 8660
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To optimize working posture while improving comfort level and cleaning efficiency. Perfex Polymer Fixed Length Handle are 100% recyclable and lightweight.

  • Super-smooth finish is pleasant to touch, even in cold conditions
  • Impervious to water, grease, gasoline, detergents, sanitizers and solvents
  • Does not rot, corrode or oxidize
  • Easy to clean
  • Color: White

Specifications

Product Type Fixed Handle
Color White
Resistant To Water, Grease, Gasoline, Detergents, Sanitizers and Solvents
For Use With (Equipment) Push Brooms
Material Polymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.