Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ TruCLEAN™ II Mopping System

Catalog No. 19042347 Shop All Perfex Corp. Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-042-347 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-042-347 Supplier Perfex™ Supplier No. 302R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

mop frame and adjustable polymer handle, a stainless-steel wringer, the 9 1/2 gal. bucket on casters, and waste containment bucket.

  • Compact double-bucket disinfecting system
  • Ideal for small area cleaning
  • System includes buckets, wringer, mop frame, and handle

Compact double-bucket system specifically designed for small area use. The waste bucket fits inside the large 9 1/2 gal. (36L) polymer bucket on casters, isolating soiled contaminants.

Specifications

Product Type Mopping System II
Color Red
Autoclavable Autoclavable
Includes Mop frame and adjustable polymer handle, a stainless steel wringer, the 9.5 gal. bucket on casters and waste containment bucket
For Use With (Application) For Small Area Cleaning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.