Learn More
Perfex™ TruCLEAN™ Mop Frame
Catalog No. 19003413 Shop All Perfex Corp. Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
19-003-413 Autoclavable
1 options
Catalog No. 19-003-413 Supplier Perfex™ Supplier No. 2238
Add to Cart
Add to Cart
Exclusive non-flipping, low-profile swivel frame design allows continual surface contact at any angle for 360° maneuverability on floors, walls, ceilings, baseboards, and any confined areas, such as corners, stairs, or under fixtures; swivel also locks in any position. Can also be used for mopping in an “S” motion to collect solid particles.
Quick-release frame locking mechanism for easy mop replacement. Constructed of high-grade stainless steel. Compatible with gamma, EtO, and autoclave sterilization.
Order Info
Adjustable Mop Handle sold separately. Contact your Fisher Safety Customer Service Representative for ordering information.
Specifications
Autoclavable Autoclavable
For Use With (Application) For floors, walls, ceilings, baseboards and any confined areas, permits mopping in an 'S' motion
Material Stainless Steel