Learn More
Perfex™ TruClean™ Yarn Mop
Catalog No. 19003412 Shop All Perfex Corp. Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
19-003-412 Autoclavable
1 options
Catalog No. 19-003-412 Supplier Perfex™ Supplier No. 2233
Add to Cart
Edge
Add to Cart
Mops are ideal for use in controlled environments or general housekeeping
  • Compatible with gamma, ETO, and autoclave sterilization
  • Inert to bacteria/chemicals
  • Constructed of synthetic yarn
  • Looped end design
Specifications
Product Type Yarn Mop
Autoclavable Autoclavable
For Use With (Application) Controlled Environments
Material Synthetic Yarn