Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Perfluoro-1-butanesulfonyl fluoride

Catalog No. 319732100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-973-2100G 100 g
31-973-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-973-2100G Supplier Sigma Aldrich Supplier No. 319732100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Perfluoro-1-butanesulfonyl fluoride (nff) reacts with alcohols, including phenols, to yield nonafluorobutanesulfonate esters (nonaflates)
  • Nonaflates can be used as electrophiles in several palladium-catalyzed cross coupling reactions and in buchwald-hartwig amination.nff can also be used to prepare aryl nonaflates by reacting with corresponding aryloxysilanes in the presence of fluoride ion

Specifications

CAS 375-72-4
Density 1.682 g/mL (at 25°C (literature))
Boiling Point 64°C (lit.)
Molecular Formula C4F10O2S
Refractive Index n20/D 1.3 (literature)
Linear Formula CF3(CF2)3 SO2F
MDL Number MFCD00007422
Quantity 100 g
Synonym NfF; Nonafluorobutanesulfonyl fluoride
Molecular Weight (g/mol) 302.09
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.