Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Perfluoro-n-octyl)phenyliodonium Trifluoromethanesulfonate, TCI America™
SDP

Catalog No. P10811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10811G Supplier TCI America Supplier No. P10811G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77758-89-5
Molecular Formula C15H5F20IO3S
Molecular Weight (g/mol) 772.135
MDL Number MFCD00135120
InChI Key LQSJCHNACSIGQK-UHFFFAOYSA-M
Synonym (Eicosafluoro-n-octyl)phenyliodonium Trifluoromethanesulfonate
PubChem CID 2760324
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl(phenyl)iodanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[I+]C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 772.13
Physical Form Crystalline Powder
Chemical Name or Material (Perfluoro-n-octyl)phenyliodonium Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.