Learn More
Medchemexpress LLC Perfluorobutylsulfonyl fluoride | 375-72-4 | 99.9% | 302.09 | 50 G
SDP
Catalog No. 5000401767
Encompass_Preferred
Perfluorobutylsulfonyl fluoride is a biological molecule. It is intended for research use only and not for sale to patients. This product is a liquid with a density of 1.774±0.06 g/cm3 and appears colorless to light yellow.
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage of pure form at -20°C for 3 years, 4°C for 2 years
  • Storage in solvent at -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-01767 Supplier Medchemexpress LLC Supplier No. HY2119050G
Perfluorobutylsulfonyl fluoride is a biological molecule. It is intended for research use only and not for sale to patients. This product is a liquid with a density of 1.774±0.06 g/cm3 and appears colorless to light yellow.
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage of pure form at -20°C for 3 years, 4°C for 2 years
  • Storage in solvent at -80°C for 6 months, -20°C for 1 month