Learn More
Cayman Chemical Perfluorooctanesulfonamide, Purity: ≥85%, CAS: 754-91-6, 100 mg
SDP
Catalog No. 502919387
Encompass_Preferred
A perfluoroalkyl substance; induces hepatic macrophage and neutrophil accumulation and fibrosis in zebrafish larvae when administered in tank water from 0.1-100 µg/L; has been found in municipal wastewater networks
Catalog No. 50-291-9387 Supplier Cayman Chemical Supplier No. 37258100 MG
A perfluoroalkyl substance; induces hepatic macrophage and neutrophil accumulation and fibrosis in zebrafish larvae when administered in tank water from 0.1-100 µg/L; has been found in municipal wastewater networks