Learn More
Medchemexpress LLC Perfluorodecalin | 306-94-5 | MFCD00010626 | 98.0% | 462.08 g/mol | C10F18 | 1 G
SDP
Catalog No. 5000243978
Encompass_Preferred
Perfluorodecalin is a fully fluorinated hydrocarbon used as an oxygen-carrying solvent and research reagent. It is applied in biomedical research (including vitreoretinal procedure models and oxygen-transport studies) and as a fluorous solvent due to its high oxygen solubility and thermal stability.
  • High oxygen solubility for oxygen-carrying and transport studies.
  • Thermally stable and non-flammable under normal laboratory conditions.
  • Used in vitreoretinal research models and related biomedical studies.
  • High research-grade purity (98.0%) suitable for experimental use.
  • Available in small laboratory quantities for benchtop experiments.
Catalog No. 50-002-43978 Supplier Medchemexpress LLC Supplier No. HY1082981G
Perfluorodecalin is a fully fluorinated hydrocarbon used as an oxygen-carrying solvent and research reagent. It is applied in biomedical research (including vitreoretinal procedure models and oxygen-transport studies) and as a fluorous solvent due to its high oxygen solubility and thermal stability.
  • High oxygen solubility for oxygen-carrying and transport studies.
  • Thermally stable and non-flammable under normal laboratory conditions.
  • Used in vitreoretinal research models and related biomedical studies.
  • High research-grade purity (98.0%) suitable for experimental use.
  • Available in small laboratory quantities for benchtop experiments.