Learn More
Medchemexpress LLC Perfluorodecyl bromide | 307-43-7 | 100.0% | 50 MG
SDP
Catalog No. 5000233042
Encompass_Preferred
Perfluorodecyl bromide (PFDB) is a high-molecular-weight, lipophilic fluorocarbon used in chemical research and formulation. It is an analogue of perflubron and is commonly used to prepare phospholipid-stabilized fluorocarbon emulsions.
  • High-molecular-weight, lipophilic fluorocarbon
  • Used to prepare phospholipid-stabilized fluorocarbon emulsions
  • Solid at room temperature; molecular weight 598.98 g/mol
  • Chemical formula C10BrF21; CAS 307-43-7
  • Available in small laboratory quantities (for example, 50 mg)
Catalog No. 50-002-33042 Supplier Medchemexpress LLC Supplier No. HY11955450MG
Perfluorodecyl bromide (PFDB) is a high-molecular-weight, lipophilic fluorocarbon used in chemical research and formulation. It is an analogue of perflubron and is commonly used to prepare phospholipid-stabilized fluorocarbon emulsions.
  • High-molecular-weight, lipophilic fluorocarbon
  • Used to prepare phospholipid-stabilized fluorocarbon emulsions
  • Solid at room temperature; molecular weight 598.98 g/mol
  • Chemical formula C10BrF21; CAS 307-43-7
  • Available in small laboratory quantities (for example, 50 mg)