Learn More
Medchemexpress LLC Perfluorododecanoic acid | 307-55-1 | MFCD00198081 | 95.0% | 614.10 g·mol⁻¹ | C12HF23O2 | 10 MG
SDP
Catalog No. 5000282042
Encompass_Preferred
Perfluorododecanoic acid is a perfluoroalkyl carboxylic acid used as a research reagent to investigate PFAS-driven biological effects such as cytotoxicity, increased reactive oxygen species, and altered mitochondrial membrane potential. It is provided as a high-purity solid for laboratory studies.
Catalog No. 50-002-82042 Supplier Medchemexpress LLC Supplier No. HYW27529510MG
Perfluorododecanoic acid is a perfluoroalkyl carboxylic acid used as a research reagent to investigate PFAS-driven biological effects such as cytotoxicity, increased reactive oxygen species, and altered mitochondrial membrane potential. It is provided as a high-purity solid for laboratory studies.