Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Perfluorododecanoic acid | 307-55-1 | MFCD00198081 | 95.0% | 614.10 g/mol | C12HF23O2 | 100 MG
SDP

Catalog No. 5000282422
Encompass_Preferred

Perfluorododecanoic acid (PFDoA) is a perfluoroalkyl carboxylic acid used as a research reagent in environmental, toxicological, and analytical studies of per- and polyfluoroalkyl substances (PFAS). It is supplied as a solid for laboratory use and is not intended for human or clinical applications.

  • 95.0% purity.
  • Molecular formula C12HF23O2.
  • Molecular weight 614.10 g/mol.
  • CAS number 307-55-1.
  • Supplied in 100 mg quantity.
  • For research use only; not for human use.
  • Suitable for PFAS environmental and toxicology studies.

Catalog No. 50-002-82422 Supplier Medchemexpress LLC Supplier No. HYW275295100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perfluorododecanoic acid (PFDoA) is a perfluoroalkyl carboxylic acid used as a research reagent in environmental, toxicological, and analytical studies of per- and polyfluoroalkyl substances (PFAS). It is supplied as a solid for laboratory use and is not intended for human or clinical applications.

  • 95.0% purity.
  • Molecular formula C12HF23O2.
  • Molecular weight 614.10 g/mol.
  • CAS number 307-55-1.
  • Supplied in 100 mg quantity.
  • For research use only; not for human use.
  • Suitable for PFAS environmental and toxicology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.