Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Perfluorododecanoic acid | 307-55-1 | MFCD00198081 | 95.0% | 614.10 g/mol | C12HF23O2 | 50 MG
SDP

Catalog No. 5000281857
Encompass_Preferred

Perfluorododecanoic acid is a perfluoroalkyl carboxylic acid used as a research reagent for studies of environmental distribution and biological effects of PFAS compounds. It is supplied as a solid for laboratory and analytical applications; consult the safety data sheet for handling and storage instructions.

  • Perfluorinated C12 carboxylic acid for PFAS research.
  • Purity 95.0% suitable for analytical methods.
  • Molecular weight 614.10 g/mol.
  • Chemical formula C12HF23O2.
  • Supplied as 50 MG solid quantity.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-81857 Supplier Medchemexpress LLC Supplier No. HYW27529550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perfluorododecanoic acid is a perfluoroalkyl carboxylic acid used as a research reagent for studies of environmental distribution and biological effects of PFAS compounds. It is supplied as a solid for laboratory and analytical applications; consult the safety data sheet for handling and storage instructions.

  • Perfluorinated C12 carboxylic acid for PFAS research.
  • Purity 95.0% suitable for analytical methods.
  • Molecular weight 614.10 g/mol.
  • Chemical formula C12HF23O2.
  • Supplied as 50 MG solid quantity.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.