Learn More
Cayman Chemical Perfluoroheptanoic Acid, Purity: ≥90%, CAS: 375-85-9, 50 g
SDP
Catalog No. 502915415
Encompass_Preferred
A perfluoroalkyl substance; prenatal exposure is positively correlated with preterm birth, primarily in male infants; positively correlated with fat mass, BMI, and waist circumference in females but negatively associated with BMI, waist circumference, and weight-to-height ratio in males
Catalog No. 50-291-5415 Supplier Cayman Chemical Supplier No. 3724750 G
A perfluoroalkyl substance; prenatal exposure is positively correlated with preterm birth, primarily in male infants; positively correlated with fat mass, BMI, and waist circumference in females but negatively associated with BMI, waist circumference, and weight-to-height ratio in males