Learn More
Medchemexpress LLC Perfluorohexanoic acid | 307-24-4 | 99.9% | 314.05 | 25 G
SDP
Catalog No. 5000402844
Encompass_Preferred
Perfluorohexanoic acid, also known as Undecafluorocaproic acid, is a biological molecule. It is designed for research applications and not for patient use. This compound exhibits an appearance of a solid below 12°C and a liquid above 14°C, presenting as colorless to off-white.
  • Biological molecule
  • For research use only
  • Appears solid below 12°C
  • Appears liquid above 14°C
  • Colorless to off-white appearance
Catalog No. 50-004-02844 Supplier Medchemexpress LLC Supplier No. 25GHY21194
Perfluorohexanoic acid, also known as Undecafluorocaproic acid, is a biological molecule. It is designed for research applications and not for patient use. This compound exhibits an appearance of a solid below 12°C and a liquid above 14°C, presenting as colorless to off-white.
  • Biological molecule
  • For research use only
  • Appears solid below 12°C
  • Appears liquid above 14°C
  • Colorless to off-white appearance