Learn More
Cayman Chemical PerfluorohexnsulfonamIde 100mg
SDP
Catalog No. 502916420
Encompass_Preferred
A perfluoroalkyl substance; has been found in surface freshwater foams and in soil from fire-fighting equipment testing sites
Catalog No. 50-291-6420 Supplier Cayman Chemical Supplier No. 38306100 MG
A perfluoroalkyl substance; has been found in surface freshwater foams and in soil from fire-fighting equipment testing sites