Learn More
Cayman Chemical Perfluorohexanesulfonamide, Purity: ≥75%, CAS: 41997-13-1, 50 mg
SDP
Catalog No. 502918075
Encompass_Preferred
A perfluoroalkyl substance; has been found in surface freshwater foams and in soil from fire-fighting equipment testing sites
Catalog No. 50-291-8075 Supplier Cayman Chemical Supplier No. 3830650 MG
A perfluoroalkyl substance; has been found in surface freshwater foams and in soil from fire-fighting equipment testing sites