Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorokerosene-L, Thermo Scientific™

Catalog No. AAL1659703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1659703 1 g
AAL1659706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1659703 Supplier Thermo Scientific Chemicals Supplier No. L1659703
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Perfluorokerosene-L is used as a standard in mass spectrometry. It is used as a solvent in the preparation of 1,3-oxazin-2-ones and oxazolidin-2-ones.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00132835

Specifications

Density 1.86
Boiling Point 70°C to 200°C
Refractive Index 1.299
Quantity 1 g
Solubility Information Immiscible with water.
Chemical Name or Material Perfluorokerosene-L
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.