Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorokerosene (Low boiling) [for Mass spectrometry], TCI America™
SDP

Catalog No. P069010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P069010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P069010G Supplier TCI America Supplier No. P069010G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00132835

Specifications

Color Colorless
Quantity 10 g
UN Number 1993
Physical Form Liquid
Chemical Name or Material Perfluorokerosene (Low boiling) [for Mass spectrometry]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.