Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Perfluorooctanoic Acid

Catalog No. 17146825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-146-825G 25 g
17-146-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-146-825G Supplier Sigma Aldrich Supplier No. 17146825G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Perfluorooctanoic Acid

Specifications

CAS 335-67-1
Melting Point 55°C to 56°C
Boiling Point 189°C (736 mmHg, lit.)
Molecular Formula C8HF15O2
Linear Formula CF3(CF2)6COOH
Quantity 25 g
MDL Number MFCD00004174
Synonym Pentadecafluorooctanoic acid; PFOA; Perfluorocaprylic acid; Perfluorooctanoic acid
Molecular Weight (g/mol) 414.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.