Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Perfluoropentanoic Acid

Catalog No. 39657525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-657-525ML 25 mL
39-657-55ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 39-657-525ML Supplier Sigma Aldrich Supplier No. 39657525ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Perfluoropentanoic acid is a short-chain perfluorocarboxylic acid (PFCA) generally used as an industrial surfactant and surface protector.

Specifications

CAS 2706-90-3
Density 1.713 g/mL at 25°C
Boiling Point 140°C
Molecular Formula C5HF9O2
Linear Formula CF3(CF2)3COOH
Refractive Index n20/D 1.294
MDL Number MFCD00040211
Quantity 25 mL
Synonym Nonafluorovaleric acid; PFPeA; Nonafluoropentanoic acid; Perfluoropentanoic acid
Molecular Weight (g/mol) 264.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.