Learn More
Cayman Chemical Perfluoropentanoic Acid, Purity: ≥85%, CAS: 2706-90-3, 10 g
SDP
Catalog No. 502915395
Encompass_Preferred
A perfluoroalkyl substance; activates PPARα in a reporter assay using COS-1 cells expressing the human receptor from 1-100 µM; topical application increases liver weight and the number of B cells, T cells, NK cells, eosinophils, and dendritic cells in the draining lymph node, decreases BUN levels, and induces epidermal hyperplasia and hyperkeratosis in mice at 5% v/v; has been found in landfill leachate and sea and river water
Catalog No. 50-291-5395 Supplier Cayman Chemical Supplier No. 3725410 G
A perfluoroalkyl substance; activates PPARα in a reporter assay using COS-1 cells expressing the human receptor from 1-100 µM; topical application increases liver weight and the number of B cells, T cells, NK cells, eosinophils, and dendritic cells in the draining lymph node, decreases BUN levels, and induces epidermal hyperplasia and hyperkeratosis in mice at 5% v/v; has been found in landfill leachate and sea and river water