Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Perfluorophenyl 3-(2-(2-(2-(1,3-dioxoisoindolin-2-yloxy)ethoxy)ethoxy)ethoxy)propanoate | 2101206-87-3 | >96.0% | 533.4 g/mol | C23H20F5NO8 | 10 MG
SDP

Catalog No. 5000202174
Encompass_Preferred

NHPI-PEG3-C2-Pfp ester is a noncleavable, trifunctional 3-unit polyethylene glycol (PEG) linker bearing an N-hydroxyphthalimide (NHPI) group and a perfluorophenyl (Pfp) activated ester. It is used for bioconjugation and in the synthesis of antibody-drug conjugates (ADCs). Intended for research use only.

  • Noncleavable 3-unit PEG linker
  • NHPI functional group for site-specific conjugation
  • Perfluorophenyl activated ester for amine coupling
  • Molecular weight about 533.4 g/mol; formula C23H20F5NO8
  • Purity greater than 96%
  • Intended for research use only, not for human or animal use

Catalog No. 50-002-02174 Supplier Medchemexpress LLC Supplier No. HY13009210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHPI-PEG3-C2-Pfp ester is a noncleavable, trifunctional 3-unit polyethylene glycol (PEG) linker bearing an N-hydroxyphthalimide (NHPI) group and a perfluorophenyl (Pfp) activated ester. It is used for bioconjugation and in the synthesis of antibody-drug conjugates (ADCs). Intended for research use only.

  • Noncleavable 3-unit PEG linker
  • NHPI functional group for site-specific conjugation
  • Perfluorophenyl activated ester for amine coupling
  • Molecular weight about 533.4 g/mol; formula C23H20F5NO8
  • Purity greater than 96%
  • Intended for research use only, not for human or animal use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.