Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Perfluoropropyl iodide

Catalog No. P1040225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1040225G 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1040225G Supplier Sigma Aldrich Supplier No. P1040225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-acetylcaprolactam has been used:
  • As initiator to investigate kinetics of anionic polymerization of α-caprolactam
  • In ring-opening polymerization of nylon 6

Specifications

CAS 754-34-7
Density 2.05 g/mL (at 25°C (literature))
Boiling Point 41°C (lit.)
Molecular Formula C3F7I
Linear Formula CF3CF2CF2I
Refractive Index n20/D 1.328 (literature)
MDL Number MFCD00001061
Quantity 250 mL
Synonym 1-Iodoperfluoropropane; Heptafluoro-1-iodopropane; Heptafluoropropyl iodide
Molecular Weight (g/mol) 295.93
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.