Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorotetradecane, 97%, Thermo Scientific™

Catalog No. AAL1684603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1684603 1 g
AAL1684606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1684603 Supplier Thermo Scientific Chemicals Supplier No. L1684603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-62-0
Molecular Formula C14F30
Molecular Weight (g/mol) 738.106
MDL Number MFCD00013571
InChI Key HYZQZWDYBKVIRI-UHFFFAOYSA-N
Synonym perfluorotetradecane, triacontafluorotetradecane, perfluorotetradecan, acmc-1ckik
PubChem CID 2776371
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-triacontafluorotetradecane
SMILES C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 103°C to 105°C
Linear Formula CF3(CF2)12CF3
Quantity 1 g
Formula Weight 738.11
Percent Purity 97%
Chemical Name or Material Perfluorotetradecane
EINECSNumber 206-206-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.