Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorotributylamine, MP Biomedicals

Catalog No. ICN207563 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN207563 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN207563 Supplier MP Biomedicals, Inc Supplier No. 0520756310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 311-89-7
Molecular Formula C12F27N
Molecular Weight (g/mol) 671.096
InChI Key RVZRBWKZFJCCIB-UHFFFAOYSA-N
Synonym perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine
PubChem CID 9397
ChEBI CHEBI:38854
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
SMILES C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Quantity 10 g
Chemical Name or Material Perfluorotributylamine
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.