Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorotripentylamine, tech. 85%, Thermo Scientific Chemicals

Catalog No. AAL1684814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1684814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1684814 Supplier Thermo Scientific Chemicals Supplier No. L1684814

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338-84-1
Molecular Formula C15F33N
Molecular Weight (g/mol) 821.119
MDL Number MFCD00042367
InChI Key AQZYBQIAUSKCCS-UHFFFAOYSA-N
Synonym perfluorotripentylamine, tris undecafluoropentyl amine, perfluorotri-n-pentylamine, perfluorotriamylamine, fluorinert fc-70, perfluoro-compound fc-70, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis undecafluoropentyl, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl pentan-1-amine, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl, fluorinert fc 70
PubChem CID 67646
IUPAC Name 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-N,N-bis(1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl)pentan-1-amine
SMILES C(C(C(N(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.94
Boiling Point 210°C to 220°C
Quantity 25 g
Formula Weight 821.12
Percent Purity 85%
Grade Technical
Chemical Name or Material Perfluorotripentylamine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 206-421-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.