Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Perfluoroundecanoic acid | 2058-94-8 | MFCD00153268 | 99.9% | 564.09 g/mol | C11HF21O2 | 500 MG
SDP

Catalog No. 5000283029
Encompass_Preferred

Perfluoroundecanoic acid is a perfluorinated 11-carbon carboxylic acid supplied for laboratory research. MedChemExpress lists it as an orally active oxidative-stress inducer that can promote macrophage M2 polarization. Use in analytical and biological studies only, and follow safety data sheet instructions for handling and storage.

  • Chemical formula C11HF21O2; molecular weight 564.09 g/mol.
  • CAS number 2058-94-8 for unambiguous identification.
  • High purity (99.85%).
  • Available in small laboratory pack sizes, including 500 mg.
  • Used as an oxidative stress inducer in biological assays.
  • Store and handle according to the safety data sheet and institutional protocols.

Catalog No. 50-002-83029 Supplier Medchemexpress LLC Supplier No. HYW142432500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perfluoroundecanoic acid is a perfluorinated 11-carbon carboxylic acid supplied for laboratory research. MedChemExpress lists it as an orally active oxidative-stress inducer that can promote macrophage M2 polarization. Use in analytical and biological studies only, and follow safety data sheet instructions for handling and storage.

  • Chemical formula C11HF21O2; molecular weight 564.09 g/mol.
  • CAS number 2058-94-8 for unambiguous identification.
  • High purity (99.85%).
  • Available in small laboratory pack sizes, including 500 mg.
  • Used as an oxidative stress inducer in biological assays.
  • Store and handle according to the safety data sheet and institutional protocols.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.