Learn More
Sklar Instruments Perforated Instrument Tray, Sklar, 16-1/2 in. x 10 in. x 2-1/2 in., Stainless Steel. The trays are used for hot or cold sterilizing, storing and transporting instruments. EA/1.
SDP
Catalog No. 501437150 Shop All Sklar Instruments Products
Encompass_Preferred
Perforated Instrument Tray, Sklar, 16-1/2 in. x 10 in. x 2-1/2 in., Stainless Steel. Sklar's Perforated Instrument Tray are seamless to prevent bacterial growth. The trays are used for hot or cold sterilizing, storing and transporting instruments for surgery. The perforated tray allows steam cleaning without moisture bild up. Dome or flat lids are available for use with tray. This instrument tray comes in a variety of sizes. The overall size is 16-1/2 inches x 10 inches x 2-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7150 Supplier Sklar Instruments Supplier No. 101894
Perforated Instrument Tray, Sklar, 16-1/2 in. x 10 in. x 2-1/2 in., Stainless Steel. Sklar's Perforated Instrument Tray are seamless to prevent bacterial growth. The trays are used for hot or cold sterilizing, storing and transporting instruments for surgery. The perforated tray allows steam cleaning without moisture bild up. Dome or flat lids are available for use with tray. This instrument tray comes in a variety of sizes. The overall size is 16-1/2 inches x 10 inches x 2-1/2 inches.