Learn More
Marlin Steel Wire Products Perforated Mesh Basket with Handles 10.57"L x 8.70"H x 6.25"H
SDP
Catalog No. NC1962209
Encompass_Preferred
Stainless steel material handling basket with a lid and hinge.Used for small material handling or washing. Rugged construction ensures basket longevity in applications such as chemical baths, heat treating, cryogenics, and food processing.
Catalog No. NC1962209 Supplier Marlin Steel Wire Products Supplier No. 000036822881
Stainless steel material handling basket with a lid and hinge.Used for small material handling or washing. Rugged construction ensures basket longevity in applications such as chemical baths, heat treating, cryogenics, and food processing.