Learn More
Marlin Steel Wire Products Perforated Mesh Basket with Handles 8.625"D x 8.25"H
SDP
Catalog No. NC1959534
Encompass_Preferred
Stainless steel material handling basket with a lid and hinge.4 mesh openings per linear inchUsed for small material handling or washing. Rugged construction ensures basket longevity in applications such as chemical baths, heat treating, cryogenics, and food processing.
Catalog No. NC1959534 Supplier Marlin Steel Wire Products Supplier No. 000036822638
Stainless steel material handling basket with a lid and hinge.4 mesh openings per linear inchUsed for small material handling or washing. Rugged construction ensures basket longevity in applications such as chemical baths, heat treating, cryogenics, and food processing.