Learn More
BINDER™ Perforated Shelf Rack
SDP
Catalog No. 502875732 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 130, KBF/KBF PRO 130
KB/KB PRO 720, KBF/KBF PRO 720
KB/KB PRO/KBF/KBF PRO 260/470
KBF/KBF PRO 1600, KB PRO 1600
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5732 KBF/KBF PRO 1600, KB PRO 1600
50-287-5580 KB/KB PRO 130, KBF/KBF PRO 130
50-287-5596 KB/KB PRO/KBF/KBF PRO 260/470
50-287-5597 KB/KB PRO 720, KBF/KBF PRO 720
50-287-5581 KB/KB PRO 1060, KBF/KBF PRO 1060
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-287-5732 Supplier BINDER™ Supplier No. 80122410
Add to Cart
Add to Cart
Perforated shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
  • Shelf rack made of stainless steel
  • Includes set of U-rails
Specifications
For Use With (Equipment) KBF/KBF PRO 1600, KB PRO 1600
Material Stainless Steel
Type Rack